1-Cyclohexylpiperazine 98%

Supplier: Ambeed
Warning

A761891-10G A761891-25G A761891-5G A761891-1G
77019-134EA 29 USD
77019-134 77019-136 77019-138 77891-496
1-Cyclohexylpiperazine 98%
1-Cyclohexylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂
MDL Number: MFCD00044809
CAS Number: 17766-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR