6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-(Boc-amino)spiro[3.3]heptane-2-carboxylate, Methyl 6-((tert-butoxycarbonyl)amino)spiro[3.3]heptane-2-carboxylate

A761759-100MG A761759-1G A761759-250MG A761759-5G
76993-120EA 22 USD
76993-120 76993-122 76993-124 76993-126
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester 95%
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₄
MDL Number: MFCD18425636
CAS Number: 170508-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR