(1-Phenyl-1H-pyrazol-4-yl)methylamine 95%

Supplier: Ambeed
Danger

Synonyms: (1-Phenyl-1H-pyrazol-4-yl)methanamine

A761646-100MG A761646-1G
77036-174EA 84.88 USD
77036-174 77036-176
(1-Phenyl-1H-pyrazol-4-yl)methylamine 95%
(1-Phenyl-1H-pyrazol-4-yl)methylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃
MW: 173.22 g/mol
MDL Number: MFCD02656664
CAS Number: 400877-10-3
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR