3-Bromo-2-(3-fluorobenzyloxy)-5-methylphenylboronic acid 95%

Supplier: Ambeed
Warning

A760643-100MG A760643-1G A760643-250MG
77019-124EA 11 USD
77019-124 77019-126 77019-128
3-Bromo-2-(3-fluorobenzyloxy)-5-methylphenylboronic acid 95%
3-Bromo-2-(3-fluorobenzyloxy)-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BBrFO₃
MDL Number: MFCD06411359
CAS Number: 849062-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR