8-Bromo-6-chloro-3-methylimidazo[1,2-a]pyrazine 95%

Supplier: Ambeed
Warning

A760598-100MG A760598-250MG A760598-50MG
76993-114EA 143 USD
76993-114 76993-116 76993-118
8-Bromo-6-chloro-3-methylimidazo[1,2-a]pyrazine 95%
8-Bromo-6-chloro-3-methylimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₇H₅BrClN₃
MDL Number: MFCD22576309
CAS Number: 84066-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR