3-(3-Nitrophenyl)-1H-pyrazol-5-amine 95%

Supplier: Ambeed
Warning

A760282-250MG A760282-5G
77019-122EA 47.75 USD
77019-122 77684-698
3-(3-Nitrophenyl)-1H-pyrazol-5-amine 95%
3-(3-Nitrophenyl)-1H-pyrazol-5-amine

Precaución: For research use only.

Formula: C₉H₈N₄O₂
MW: 204.18 g/mol
MDL Number: MFCD05261908
CAS Number: 2765570-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR