tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 98%

Supplier: Ambeed
Warning

A759977-100MG A759977-250MG
76993-108EA 28 USD
76993-108 77891-494
tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 98%
tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD17016673
CAS Number: 227940-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR