3,5-Dibromo-1-methyl-1H-pyrazole 98%
Supplier: Ambeed
|
Warning
|
A759525-250MG
A759525-5G
A759525-1G
76993-098EA
51.99
USD
76993-098
76993-100
77684-696
3,5-Dibromo-1-methyl-1H-pyrazole 98%
3,5-Dibromo-1-methyl-1H-pyrazole
Precaución: For research use only.
Formula:
C₄H₄Br₂N₂ MW: 239.89 g/mol |
MDL Number:
MFCD28399417 CAS Number: 1361019-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...