1-(Methylsulfonyl)-4-piperidinecarboxylic acid 95%

Supplier: Ambeed
Warning

A759235-10G A759235-250MG A759235-5G A759235-1G
77036-162EA 111 USD
77036-162 77036-164 77036-166 77873-204
1-(Methylsulfonyl)-4-piperidinecarboxylic acid 95%
1-(Methylsulphonyl)-4-piperidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄S
MDL Number: MFCD03821886
CAS Number: 280772-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR