N-[2-(Aminomethyl)phenyl]-N,N-dimethylamine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)-N,N-dimethylaniline

A758670-1G A758670-250MG A758670-5G
77006-312EA 57.29 USD
77006-312 77006-314 77006-316
N-[2-(Aminomethyl)phenyl]-N,N-dimethylamine 98%
N-[2-(Aminomethyl)phenyl]-N,N-dimethylamine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MW: 150.23 g/mol
MDL Number: MFCD04037893
CAS Number: 57678-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR