Boc-Lys(boc)-ONP 98%

Supplier: Ambeed
Warning

A757992-1G A757992-25G A757992-5G
77006-306EA 14 USD
77006-306 77006-308 77006-310
Boc-Lys(boc)-ONP 98%
Boc-Lys(boc)-ONP

Precaución: for Research Use Only

Formula: C₂₂H₃₃N₃O₈
MDL Number: MFCD00076961
CAS Number: 2592-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR