1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-1,2,3,6-tetrahydropyridine, N-BOC protected, tert-Butyl 5-hydroxy-5,6-dihydropyridine-1(2H)-carboxylate

A757877-100MG A757877-1G A757877-250MG
77019-110EA 47.75 USD
77019-110 77684-694 77684-695
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine 97%
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine

Precaución: For research use only.

Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD04972246
CAS Number: 224779-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR