(3-(((Tetrahydrofuran-2-yl)methyl)carbamoyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A757849-100MG A757849-1G A757849-250MG
76993-086EA 37 USD
76993-086 76993-088 76993-090
(3-(((Tetrahydrofuran-2-yl)methyl)carbamoyl)phenyl)boronic acid 98%
(3-(((Tetrahydrofuran-2-yl)methyl)carbamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆BNO₄
MDL Number: MFCD12546614
CAS Number: 1218790-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR