4-Fluoro-2-nitrobenzamide 97%

Supplier: Ambeed
Warning

A757361-10G A757361-1G A757361-250MG A757361-5G
76993-070EA 337.37 USD
76993-070 76993-072 76993-074 76993-076
4-Fluoro-2-nitrobenzamide 97%
4-Fluoro-2-nitrobenzamide

Precaución: for Research Use Only

Formula: C₇H₅FN₂O₃
MW: 184.1246 g/mol
MDL Number: MFCD04108565
CAS Number: 106754-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR