(3S,5S)-3,5-Dimethylmorpholine 98%

Supplier: Ambeed
Danger

A757325-1G A757325-250MG A757325-100MG
77036-154EA 267.35 USD
77036-154 77036-156 77670-038
(3S,5S)-3,5-Dimethylmorpholine 98%
(3S,5S)-3,5-Dimethylmorpholine

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.17 g/mol
MDL Number: MFCD14586370
CAS Number: 154634-96-5
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR