1-Octanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
n-Octanol
A757153-100G
A757153-500G
A757153-1000G
77006-298EA
6.37
USD
77006-298
77006-300
77569-446
1-Octanol 98%
1-Octanol
Precaución: For research use only.
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...