3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Difluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A757098-100MG A757098-1G A757098-250MG
77019-096EA 37.14 USD
77019-096 77019-098 77019-100
3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester 95%
3,5-Difluoro-2-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
MDL Number: MFCD07781207
CAS Number: 1073354-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR