2-Fluoro-3-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A757023-1G A757023-250MG
76993-066EA 60 USD
76993-066 76993-068
2-Fluoro-3-iodobenzonitrile 95%
2-Fluoro-3-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
MW: 247.008 g/mol
MDL Number: MFCD18396938
CAS Number: 211943-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR