3-(1H-Imidazol-1-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Formylphenyl)-1H-imidazole, 3-(1-Imidazolyl)benzaldehyde

A756355-10G A756355-1G A756355-250MG A756355-5G
77006-292EA 96.55 USD
77006-292 77006-294 77006-296 77674-074
3-(1H-Imidazol-1-yl)benzaldehyde 97%
3-(1H-Imidazol-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.183 g/mol
MDL Number: MFCD09836178
CAS Number: 127404-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR