7-Bromo-5-chlorobenzofuran 95%

Supplier: Ambeed
Warning

A756278-1G A756278-250MG
76993-062EA 114.58 USD
76993-062 76993-064
7-Bromo-5-chlorobenzofuran 95%
7-Bromo-5-chlorobenzofuran

Precaución: for Research Use Only

Formula: C₈H₄BrClO
MW: 231.47 g/mol
MDL Number: MFCD09056789
CAS Number: 286836-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR