Undecaethylene glycol 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A755793-1G A755793-250MG A755793-5G A755793-100MG
77019-084EA 30 USD
77019-084 77019-086 77019-088 77684-690
Undecaethylene glycol 98%, Ambeed.Inc
Undecaethylene glycol

Precaución: For research use only.

Formula: C₂₂H₄₆O₁₂
MDL Number: MFCD26793839
CAS Number: 6809-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR