5-(3-Nitrophenyl)isoxazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A755723-1G A755723-250MG A755723-5G
77019-078EA 80.63 USD
77019-078 77019-080 77019-082
5-(3-Nitrophenyl)isoxazole-3-carboxylic acid 97%
5-(3-Nitrophenyl)isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₅
MW: 234.17 g/mol
MDL Number: MFCD11217082
CAS Number: 199601-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR