Nonaethylene glycol monomethyl ether 98%

Supplier: Ambeed
Warning

Synonyms: 2,5,8,11,14,17,20,23,26-Nonaoxaoctacosan-28-ol, mPEG9-Alcohol

A755540-1G A755540-250MG A755540-25G A755540-5G A755540-100G
77019-072EA 22 USD
77019-072 77019-074 77019-076 77614-330 77891-488
Nonaethylene glycol monomethyl ether 98%
Nonaethylene glycol monomethyl ether

Precaución: for Research Use Only

Formula: C₁₉H₄₀O₁₀
MW: 428.52 g/mol
MDL Number: MFCD06797148
CAS Number: 6048-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR