(2-Methyl-3-nitrophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A755528-100G A755528-10G A755528-1G A755528-25G A755528-5G
77006-282EA 94 USD
77006-282 77006-284 77006-286 77006-288 77006-290
(2-Methyl-3-nitrophenyl)acetic acid 97%
(2-Methyl-3-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD07782132
CAS Number: 23876-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR