2-Acetylthiophene-3-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetyl-3-thiopheneboronic acid, 2-Acetyl-3-thienylboronic acid, (2-Acetylthiophen-3-yl)boronic acid

A755066-100MG A755066-1G A755066-250MG A755066-5G
76993-052EA 23.34 USD
76993-052 76993-054 76993-056 76993-058
2-Acetylthiophene-3-boronic acid 98%
2-Acetylthiophene-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₃S
MW: 169.9940 g/mol
MDL Number: MFCD01318932
CAS Number: 36155-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR