N-Cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A754743-1G A754743-250MG A754743-5G
76993-046EA 84.88 USD
76993-046 76993-048 76993-050
N-Cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide 98%
N-Cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₄S
MW: 323.22 g/mol
MDL Number: MFCD06657824
CAS Number: 914610-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR