trans-4-[Boc(methyl)amino]cyclohexanol 97%
Supplier: Ambeed
|
Warning
|
A754450-250MG
A754450-1G
76993-044EA
32.89
USD
76993-044
77684-687
trans-4-[Boc(methyl)amino]cyclohexanol 97%
trans-4-[Boc(methyl)amino]cyclohexanol
Precaución: For research use only.
Formula:
C₁₂H₂₃NO₃ MW: 229.31 g/mol |
MDL Number:
MFCD23106043 CAS Number: 400899-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...