Benzo[b]thiophen-3-amine hydrochloride 96%

Supplier: Ambeed
Warning

A752868-1G A752868-250MG
77006-266EA 257.8 USD
77006-266 77006-268
Benzo[b]thiophen-3-amine hydrochloride 96%
Benzo[b]thiophen-3-amine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₈ClNS
MW: 185.67 g/mol
MDL Number: MFCD18157696
CAS Number: 3394-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR