Methyl 2-(3-methylpyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A752846-100MG A752846-1G A752846-250MG
77019-040EA 54 USD
77019-040 77019-042 77019-044
Methyl 2-(3-methylpyridin-2-yl)acetate 95%

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD18802818
CAS Number: 1092477-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR