Ethyl Imidazo[1,2-a]pyridine-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl imidazo[1,2-a]pyridine-5-carboxylate

A752738-100MG A752738-1G A752738-250MG
77036-136EA 47.75 USD
77036-136 77036-138 77036-140
Ethyl Imidazo[1,2-a]pyridine-5-carboxylate 95%
Ethyl Imidazo[1,2-a]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
MDL Number: MFCD13176744
CAS Number: 177485-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR