Diisopropyl-3,3-dimethoxycyclobutane-1,1-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Diisopropyl 3,3-dimethoxycyclobutane-1,1-dicarboxylate

A751041-100G A751041-10G A751041-1G A751041-25G A751041-500G A751041-5G
77036-124EA 143.23 USD
77036-124 77036-126 77036-128 77036-130 77036-132 77036-134
Diisopropyl-3,3-dimethoxycyclobutane-1,1-dicarboxylate 95%
Diisopropyl-3,3-dimethoxycyclobutane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₄O₆
MW: 288.34 g/mol
MDL Number: MFCD06742779
CAS Number: 115118-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR