3'-(Trifluoromethyl)[1,1'-biphenyl]-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 3'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine

A750975-1G A750975-250MG
77006-254EA 164.44 USD
77006-254 77006-256
3'-(Trifluoromethyl)[1,1'-biphenyl]-3-amine 95%
3'-(Trifluoromethyl)[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃N
MW: 237.23 g/mol
MDL Number: MFCD03424670
CAS Number: 400749-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR