5-Fluoro-3-methylbenzofuran-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A750227-100MG A750227-250MG A750227-1G
77006-250EA 17 USD
77006-250 77006-252 77674-064
5-Fluoro-3-methylbenzofuran-2-carboxylic acid 98%
5-Fluoro-3-methylbenzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇FO₃
MDL Number: MFCD04448734
CAS Number: 81718-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR