6-Phenyl-2H-pyran-4(3H)-one 95%

Supplier: Ambeed
Warning

A749139-100MG A749139-250MG
77019-008EA 53 USD
77019-008 77019-010
6-Phenyl-2H-pyran-4(3H)-one 95%
6-Phenyl-2H-pyran-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MDL Number: MFCD24691703
CAS Number: 5198-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR