9,9-Di-n-octylfluorene-2,7-diboronic acid 98%

Supplier: Ambeed
Warning

A748442-100MG A748442-1G A748442-250MG A748442-5G
77036-114EA 7 USD
77036-114 77036-116 77036-118 77036-120
9,9-Di-n-octylfluorene-2,7-diboronic acid 98%
9,9-Di-n-octylfluorene-2,7-diboronic acid

Precaución: for Research Use Only

Formula: C₂₉H₄₄B₂O₄
MDL Number: MFCD03701608
CAS Number: 258865-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR