1,2,3,4-Tetrahydronaphthalene 95%

Supplier: Ambeed
Danger

A748049-25G A748049-100G
77006-238EA 9.55 USD
77006-238 77573-388
1,2,3,4-Tetrahydronaphthalene 95%
1,2,3,4-Tetrahydronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₁₂
MW: 132.21 g/mol
MDL Number: MFCD00001733
CAS Number: 119-64-2
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR