2-Chloro-3-(difluoromethoxy)pyridine 95%

Supplier: Ambeed
Warning

A748020-100MG A748020-1G
77019-004EA 20.16 USD
77019-004 77019-006
2-Chloro-3-(difluoromethoxy)pyridine 95%
2-Chloro-3-(difluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClF₂NO
MW: 179.55 g/mol
MDL Number: MFCD13185847
CAS Number: 1206977-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR