2-Chloro-3-methyl-1,8-naphthyridine 95%

Supplier: Ambeed
Warning

A747215-100MG A747215-250MG
77036-112EA 166.57 USD
77036-112 77670-016
2-Chloro-3-methyl-1,8-naphthyridine 95%
2-Chloro-3-methyl-1,8-naphthyridine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MW: 178.61 g/mol
MDL Number: MFCD13193570
CAS Number: 1245649-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR