2,2-Dimethyl-1,3-benzodioxole 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-(Isopropylidenedioxy)benzene, 2,2-Dimethylbenzo[d][1,3]dioxole

A747180-1G A747180-250MG A747180-5G
76993-022EA 32.89 USD
76993-022 76993-024 77614-300
2,2-Dimethyl-1,3-benzodioxole 97%
2,2-Dimethyl-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD08061594
CAS Number: 14005-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR