2-Bromothieno[3,2-c]pyridin-4-ol 97%
Supplier: Ambeed
|
Warning
|
A747106-100MG
A747106-1G
A747106-250MG
A747106-5G
77036-104EA
76.39
USD
77036-104
77036-106
77036-108
77036-110
2-Bromothieno[3,2-c]pyridin-4-ol 97%
2-Bromothieno[3,2-c]pyridin-4-ol
Precaución: for Research Use Only
Formula:
C₇H₄BrNOS MW: 230.08 g/mol |
MDL Number:
MFCD19687059 CAS Number: 28948-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...