(S)-(-)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Xyl-SDP, (S)-7,7'-Bis(bis(3,5-dimethylphenyl)phosphaneyl)-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]

A747099-100MG A747099-1G A747099-250MG A747099-50MG
77036-096EA 376.62 USD
77036-096 77036-098 77036-100 77036-102
(S)-(-)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane 98%
(S)-(-)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane

Precaución: for Research Use Only

Formula: C₄₉H₅₀P₂
MW: 700.8688 g/mol
MDL Number: MFCD08459343
CAS Number: 528521-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR