4-(2-Bromoethoxy)benzaldehyde 95%

Supplier: Ambeed
Warning

A746944-100MG A746944-1G A746944-250MG A746944-5G
77006-230EA 13 USD
77006-230 77006-232 77006-234 77006-236
4-(2-Bromoethoxy)benzaldehyde 95%
4-(2-Bromoethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD00021074
CAS Number: 52191-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR