5-Amino-2,4-di-tert-butylphenyl methyl carbonate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A746564-100MG A746564-250MG A746564-1G
77018-996EA 30 USD
77018-996 77018-998 77684-674
5-Amino-2,4-di-tert-butylphenyl methyl carbonate ≥95%, Ambeed.Inc
5-Amino-2,4-di-tert-butylphenyl methyl carbonate

Precaución: For research use only.

Formula: C₁₆H₂₅NO₃
MDL Number: MFCD21606649
CAS Number: 1182822-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR