Benzyl 2-azabicyclo[2.2.1]hept-5-ene-2-carboxylate 95%

Supplier: Ambeed
Warning

A745912-100MG A745912-1G A745912-250MG
76992-998EA 45 USD
76992-998 76993-000 76993-002
Benzyl 2-azabicyclo[2.2.1]hept-5-ene-2-carboxylate 95%
Benzyl 2-azabicyclo[2.2.1]hept-5-ene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₂
MDL Number: MFCD24038879
CAS Number: 140927-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR