2-Amino-5-methoxybenzenesulfonic acid 98%

Supplier: Ambeed
Warning

A745839-25G A745839-500G
77006-226EA 19.1 USD
77006-226 77006-228
2-Amino-5-methoxybenzenesulfonic acid 98%
2-Amino-5-methoxybenzenesulphonic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₄S
MW: 203.2157 g/mol
MDL Number: MFCD00035804
CAS Number: 13244-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR