1H-Pyrazole-5-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrazol-5-carbaldehyde, 5-Formyl-1H-pyrazole

A745815-100MG A745815-1G A745815-250MG A745815-25G
77036-088EA 23.34 USD
77036-088 77036-090 77036-092 77036-094
1H-Pyrazole-5-carbaldehyde 95%
1H-Pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 96.087 g/mol
MDL Number: MFCD06657574
CAS Number: 948552-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR