N-(tert-Butyl)-3-((2-chloro-5-methylpyrimidin-4-yl)amino)benzenesulfonamide 95%

Supplier: Ambeed
Warning

A745808-100MG A745808-1G A745808-250MG A745808-5G
77036-080EA 83.82 USD
77036-080 77036-082 77036-084 77036-086
N-(tert-Butyl)-3-((2-chloro-5-methylpyrimidin-4-yl)amino)benzenesulfonamide 95%
N-(tert-Butyl)-3-((2-chloro-5-methylpyrimidin-4-yl)amino)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₅H₁₉ClN₄O₂S
MW: 354.85 g/mol
MDL Number: MFCD16619372
CAS Number: 936092-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR