3-Phenoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Phenoxyphenyl)boronic acid, 3-Phenoxybenzeneboronic acid

A744885-250MG A744885-1G A744885-5G
77006-224EA 21 USD
77006-224 77573-382 77882-631
3-Phenoxyphenylboronic acid 98%
3-Phenoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁BO₃
MDL Number: MFCD07780272
CAS Number: 221006-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR