1-(3-Chloro-5-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone 95%

Supplier: Ambeed
Warning

A744788-100MG A744788-1G A744788-250MG A744788-5G
76992-992EA 10.61 USD
76992-992 76992-994 76992-996 77614-284
1-(3-Chloro-5-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone 95%
1-(3-Chloro-5-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₃ClF₆O
MW: 276.56 g/mol
MDL Number: MFCD19440960
CAS Number: 1125812-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR