3-Bromo-4-(trifluoromethyl)benzyl bromide 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Bromo-4-(bromomethyl)-1-(trifluoromethyl)benzene, 2-Bromo-4-(bromomethyl)benzotrifluoride
A744018-1G
A744018-250MG
77036-068EA
151
USD
77036-068
77036-070
3-Bromo-4-(trifluoromethyl)benzyl bromide 97%
3-Bromo-4-(trifluoromethyl)benzyl bromide
Precaución: for Research Use Only
Formula:
C₈H₅Br₂F₃ MW: 317.929 g/mol |
MDL Number:
MFCD13185413 CAS Number: 372120-77-9 UN: 3265 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...